Would you like to log in again?
VERSALON® Nonwoven Sponges, Covidien
Supplier: Covidien
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VERSALON®
9024 9022 8044 9023
82004-826CS 305.17 USD InStock
82004-826 82004-828 82004-818 82013-218
VERSALON® Nonwoven Sponges, Covidien
First Aid Supplies Wound Dressings
These economical nonwoven sponges are ideal for applying ointments, prepping, wiping needles, cleaning slides, and more.
  • They are made of a rayon/polyester blend
  • Highly absorbent
  • Low-linting
Ordering information: Nonsterile sponges are bulk packaged. Sterile sponges are packaged two to a peel-back package.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty