Would you like to log in again?
Heidolph® Unimax Orbital Shakers
Supplier: Heidolph
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Heidolph®
036130180 036130520 036130200CA 036130180CA 036130520CA 036130200
63400-138EA 8045.86 USD InStock
63400-138 CA11009-592 82004-960 82004-962 CA11009-556 CA11009-554
Heidolph® Unimax Orbital Shakers
Shakers and Mixers Orbital Shakers
These orbital shakers provide the swirling action needed for the cultivation of microorganisms and cell or tissue cultures.
The Unimax package combines the model 1010 orbital shaker with an incubator for complete shaking, mixing, and heating applications. Also included is a high-profile incubator hood with a total height of 267mm (101/2"). Hoods are manufactured from transparent, non-fogging PETG material for easy viewing. Packages are available in 115V electrical capacity.
The separatory funnel attachment (82004-980) holds up to four separatory funnels (50 ml, conical) or separatory flasks (100 ml, conical). Tension roller attachment (82004-976 and -986) feature two rollers to facilitate the shaking of Erlenmeyer flasks and a piece that enables attachment to the perforated platform. The basic tension roller frame (82004-984) contains all the necessary pieces to construct a frame, but no tension rollers. Clamps for Erlenmeyer flasks and separatory funnels consist of the clamp and a piece that enables attachment to the perforated platform.
Designed for use with culture plates and Erlenmeyer flasks (≤ 1000 ml). Shakers are designed for continuous use and feature a sparkless, maintenance-free motor. Units feature timed or continuous operation modes. Digital process timer allows for unattended operation up to 999 minutes and sends an acoustic signal upon completion. Enclosed housing ensures virtually no heat transfer from motor to platform, and the unit will self-reset to protect against overheating. Units also have analog dial control of vibration frequency. The maintenance-free, balanced, sealed drive system is isolated from spills, which makes the units ideal for use in cold rooms.
Certifications: Unimax 1010 meets the standards of protection class IP 40 (DIN 40050). Unimax 2010 meets the standards of protection class IP 20 (DIN 40050).
Ordering information: Manufacturer's three-year warranty. Units include a nonskid rubber mat. Unimax 1010 includes a 29x25.8 cm (113/8x101/8") platform; Unimax 2010 includes a 39x34 cm (153/8x133/8") platform. The model 1000 incubator (82005-012) may be ordered separately, or included in a Unimax package to create a system with reproducible temperature control. For 230V units, contact your VWR sales representative.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty