Would you like to log in again?
SUPPORT ROD F/THERMETR + SENSR
Supplier: Heidolph NA, LLC
Support Rod for Contact Thermometer^ 42.5Lx1.3dia.cm. Use with contact thermometers (82005-049, 82005-050) and temperature sensors (82005-060, 82005-061, 82005-062, 82005-063). Requires clamp 82005-058.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SUPPORT ROD F/THERMETR + SENSR 82005-057 036300880 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty