Would you like to log in again?
Puritan® Oversized Rayon Swab, Proctoscopic Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
816 816
82005-438CS 365.89 USD InStock
82005-438 CA82005-438
Puritan® Oversized Rayon Swab, Proctoscopic Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
This swab has an extra-absorbent rayon tip
Autoclavable to 121°C (250°F). Made in USA.
The long polypropylene handle make it ideal for collection in the throat, and for gynecological or proctological exams.
Certifications: CE marked.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty