Would you like to log in again?
MTS 2/4 Digital Microtiter Shaker, IKA® Works
Supplier: IKA
3208001
82006-096EA 3055.62 USD InStock
82006-096
MTS 2/4 Digital Microtiter Shaker, IKA® Works
Shakers and Mixers Orbital Shakers
Microplate shaker with electronic control keeps the set speed constant via rated/actual speed comparison. Supplied with attachment but without microtiter plate.
  • Ideal for shaking two to four microtiter plates
  • Electronic speed control
Shaker features a digital timer, an alarm to indicate when set time has exprired, and platform.
Ideal for use under controlled conditions, can be used at 5 to 40 °C, in an incubator or CO₂ incubator (up to 80% RH).
Certifications: IP protection class according to DIN EN 60529: IP 21.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty