Would you like to log in again?
VWR® Nutating Mixer
Supplier: VWR
VWR®
S0500-VWR
82007-202EA 461.61 USD InStock
82007-202
VWR® Nutating Mixer
Shakers and Mixers Rotating/Rocking Shakers
The three-dimensional shaking motion of this mixer is gentle yet efficient, making it ideal for mixing samples in blood and centrifuge tubes.
  • Three-dimensional motion
  • Provides thorough yet gentle mixing
  • Ideal for blood tubes and centrifuge tubes
Mixer features a steel platform. Permanently lubricated bearings on moving parts provide quiet, maintenance-free operation. Speed and tilt angle are fixed to provide the optimal level of agitation for mixing samples in small containers without foaming. Mixer can be used in a cold room. A small footprint allows the mixer to be placed on a crowded bench.
Ordering information: Supplied with a dimpled rubber mat for firmly holding blood collection tubes, microtubes, or 15 and 50 ml centrifuge tubes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty