Would you like to log in again?
VWR® Magnetized Pipette Rack
Supplier: VWR
VWR®
HS20612MV
82007-220EA 272.35 USD InStock
82007-220
VWR® Magnetized Pipette Rack
Pipette Stands
Durable blue ABS plastic storage rack features circular magnets for secure mounting to metal fume hoods, refrigerators, file cabinets, and more.
  • Durable construction
  • Blue color
  • Magnetized
Four angled compartments accommodate 0.1 mL to 25 mL pipettes. Includes two movable attachments to hold pipetting devices. Attachment dimensions: 10.5Lx3Hcm (41/8x11/4").
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty