Would you like to log in again?
Replacement Needles for Agilent ALS GC Syringes, Hamilton Company
Supplier: Hamilton
7786-02 7786-01
82007-244PK 122.4 USD InStock
82007-244 82007-246
Replacement Needles for Agilent ALS GC Syringes, Hamilton Company
Syringe Needles
Hamilton Company offers the most comprehensive selection of standard and custom needles in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. These replacement needles are designed to work with small hub removable needle syringe for Agilent 7673, 7683, 7693 and 6850 automatic liquid sampler (ALS) gas chromatography (GC) autosamplers.
Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle Point Styles:
AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration.
Available gauges: 23, 26
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty