Synonyms:
Dihydrogen oxide
SH30529.01
SH30529.LS
SH30529.03
SH30529.01
SH30529.LS
SH30529.02
SH30529.FS
SH30529.02
SH30529.03
SH30529.FS
82007-330CS
186.72
USD
InStock
CA82007-330
82007-330
82007-326
16750-098
CA82007-328
CA82007-326
82007-328
CA16750-098
CA16750-114
16750-114
Water, HyClone™ HyPure, Cell Culture Grade
Water
Eliminate endotoxin concerns with endotoxin-free HyClone Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.
- Undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP)
- Meets < 0.005 EU/mL endotoxin by turbidimetric assay
- Manufactured in our ISO 9001 certified facilities
HyClone HyPure Cell Culture Grade Water can be used in multiple cell culture applications. With endotoxin levels as low as < 0.005 EU/mL, HyPure Cell Culture Grade Water supports healthy cell growth and consistent experimental results.
Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).