Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, HyClone™ HyPure, Molecular Biology Grade

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Dihydrogen oxide

SH30538.FS SH30538.01 SH30538.FS SH30538.02 SH30538.LS SH30538.03 SH30538.01 SH30538.03 SH30538.LS SH30538.02
82007-332EA 18.47 USD InStock
82007-332 CA16750-128 82007-336 CA82007-334 82007-334 CA82007-332 CA82007-336 16750-104 CA16750-104 16750-128
Water, HyClone™ HyPure, Molecular Biology Grade
Water
Nuclease-free HyClone Molecular Biology-Grade Water, containing no DEPC or other carcinogenic chemicals is used for critical applications involving nucleotides such as RNA and DNA.

  • Significantly low levels of endotoxins
  • Certified to have no detectable activity in DNase, RNase, and protease assays
  • Maximum allowable limit of endotoxin is 0.05 EU/mL

HyClone HyPure Molecular Biology Grade Water purification process includes continuous deionization, reverse osmosis, ultraviolet light treatment, distillation, and 0.1 µm filtration.

Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).filtration


Formula: H₂O
MW: 18.02 g/mol
Melting Pt: 0 °C
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now
Specification Test Results
Endotoxin Concentration ≤ 0.05 EU/mL
Sterilisation 0.1 µm sterile filtered
pH 5.0 to 7.0
Storage 2° to 30° C

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty