Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyClone® Bovine Calf Serum, U.S. Origin

Supplier: HyClone products (Cytiva)
HyClone™
SH30073.04HI SH30073.03HI 3007304IR25-40 SH30073.02HI 3007303IR25-40 3007304IR25-40 SH30073.02 SH30073.03IR SH30073.02HI 3007303IR25-40 SH30073.04IR SH30073.03 SH30073.04HI SH30073.02IR SH30073.02 SH30073.02IR SH30073.03HI SH30073.03 SH30073.04 SH30073.04
82007-424EA 131.25 USD InStock
82007-424 CA76324-872 CA76534-652 76324-872 76534-652 CA82007-424 CA76237-688 76237-688 CA76534-626 76534-626 CA76237-694 76237-696 76237-694 76237-692 CA76237-690 76237-690 CA16777-206 CA16777-204 16777-206 16777-204
HyClone® Bovine Calf Serum, U.S. Origin
Sera (Serum)
HyClone Bovine Calf Serum, U.S, can be used as an effective alternative to foetal bovine serum (FBS).

  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing
  • Endotoxin Concentration ≤ 10 EU/mL
  • Storage Temperature is –10 °C

Origin is carefully collected, processed and filtered in U.S. abbatoirs. Ensures excellent culture results without safety concerns.

Certifications: Fully traceable and conforms to all USDA regulations.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty