Would you like to log in again?
WYPALL® L40 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WYPALL®
05796 03046 05701 05740 05600 05796 05790 05740 05776 05600 05007 05790 05701 05007 05776 03046
82007-774CS 145.76 USD InStock
CA82007-774 CA21908-564 82007-774 14224-028 21908-000 CA21908-000 CA14224-028 89002-990 CA89011-792 89002-998 89011-792 89000-692 CA89000-692 CA89002-998 89000-684 CA89000-684
WYPALL® L40 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Disposable Wipes and Towels Wipes for General Applications
A lighter-weight wiper at a great everyday price.
  • 100% virgin fiber
  • Extra low linting, anti-static
  • Wipe clean without scratching or leaving any residue
  • For wiping surfaces, parts, instruments, and lens cleaning
Disposable wipers feature double re-creped (DRC) base sheet technology with high surface softness to absorb large liquid spills and leaks. The reinforced grid provides strength for wet-surface wiping and cleaning. Ideal for routine industrial cleaning and maintenance as well as for removing soils and liquids from face and hands.
Handy, space-saving POP-UP™ dispenser box dispenses wipers one at a time, to reduce waste.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty