Would you like to log in again?
Mini Magnifier, Heathrow Scientific®
Supplier: Heathrow Scientific
HS15092D
82008-002EA 171.14 USD InStock
82008-002
Mini Magnifier, Heathrow Scientific®
Magnifiers
The Mini Magnifier enlarges objects for easier viewing
The magnifier is equipped with a swiveling, molded lens that provides 1.75X magnification. The magnifier stand is 207 mm high.
Ordering information: Mini LED light box must be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty