Would you like to log in again?
Removable Small Hub Needles for Syringes 2.5 μl to 10 ml, Hamilton Company
Supplier: Hamilton
7803-05 7803-01 7784-06 7762-03 8646-03 7784-07 7784-05 7768-02 7768-01 7758-01 7762-01 7770-01 7762-04 7758-03 7758-04 7758-02 7784-08 7762-06 7762-02 7762-05 7770-02
82010-250PK 264.35 USD InStock
82010-250 82010-252 82010-230 82010-240 82010-236 82010-258 82010-246 82010-224 82007-208 82010-238 82010-248 82010-226 82010-254 82010-232 82010-242 82010-234 82010-244 10746-556 82010-228 89221-012 89210-098
Removable Small Hub Needles for Syringes 2.5 μl to 10 ml, Hamilton Company
Syringe Needles
Hamilton Company offers the most comprehensive selection of standard and custom needles in the industry. Each is manufactured to achieve the highest level of accuracy and precision possible.
Removable Needle (RN)- The needles are removable and are a Hamilton-specific design.
Needle Point Styles (Custom gauges, lengths and point styles available):
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-10.
Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34-10.
Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-22.
The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Ordering information: The custom made needles are available in any length from 9.5 mm (3/8") to 305 mm (12"). Available custom tip styles are 2, 3 and 4.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty