Would you like to log in again?
Standard Metal Hub Luer Lock Needles
Supplier: Hamilton
91033 90029 7730-02 91029 91014 7729-12 90023 7729-09 90051 7748-08 7729-04 7729-08 7729-06 7749-03 7729-03 7729-02 90025 90027 91032 90018 7730-01 7729-05 90015 91022 91016 90053 91012 7730-04 91019 91015 90011 90038 90019 91018 7729-07 7730-05 90012 91011 7729-01 7729-13 90014 90016 7729-14 90052 91010 91030 91031 90028 90020 7729-11 90032 91028 90013 90017 7729-10 7730-03 91017 90010 90021 91013
82010-360PK 202.71 USD InStock
82010-360 89220-914 72310-218 89220-912 89220-910 74980-012 74980-010 82010-364 82010-366 72310-180 10217-432 89220-908 72310-268 72310-266 89220-906 10217-430 10217-436 72310-248 89220-968 10217-434 10217-438 72310-208 89210-104 60377-969 72310-250 89221-156 72310-252 89221-158 89220-918 10217-440 89220-916 72310-238 82010-460 10217-406 10217-404 89220-898 89220-896 82010-348 10217-408 89221-546 72310-160 72310-242 72310-240 10217-410 72310-246 72310-200 72310-244 89220-948 82010-370 89220-904 10217-414 89220-902 10217-412 89220-900 10217-418 10217-416 82010-376 72310-270 60378-065 10217-420
Standard Metal Hub Luer Lock Needles
Syringe Needles
Hamilton company offers the most comprehensive selection of standard and custom needles in the industry.
  • Needles are made of 304 stainless steel
Each is manufactured to achieve the highest level of accuracy and precision possible. Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors.
PTFE Luer Lock (TLL)- This termination has a PTFE, male luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters and dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25 ml and greater syringes. Repeated autoclaving will shorten syringe life.
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34 to 10. Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34 to 10. Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26 to 10.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty