Would you like to log in again?
HyClone™ CDM4CHO, CHO Cell Culture Platform Liquid Media, Cytiva
Supplier: HyClone products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyClone™
SH30557.02 SH30557.02 SH30556.01 SH30558.02 SH30558.02 SH30556.01
82013-300EA 118.57 USD InStock
CA82013-300 82013-300 82013-294 76635-876 CA82013-294 CA76635-876
HyClone™ CDM4CHO, CHO Cell Culture Platform Liquid Media, Cytiva
Cell Culture Media
HyQ® CDM4CHO™ is a chemically-defined protein-free medium containing no animal derived components. It is designed to increase the process yields for the industrial manufacture of recombinant proteins using a variety of CHO cell clones. HyQ® CDM4CHO™ has been successfully tested in various culture systems, including T-flasks, shaker flasks and bioreactors. HyQ® CDM4CHO™ contains Pluronic® F68 and L-glutamine, and does not contain phenol red.
  • Metabolically designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies
  • All components fully traceable
  • Manufactured under cGMP
HyClone CDM4CHO contains Pluronic F68 and L-glutamine, and does not contain phenol red. It is also available without L-glutamine to support the GS gene expression system.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty