Would you like to log in again?
Labtainer™ BioProcess Containers, Thermo Scientific
Supplier: Thermo Scientific
Labtainer™
SH30658.14 SH30662.12 SH30657.13 SH30658.13 SH30657.16 SH30662.15 SH30662.16 SH30657.15 SH30657.11 SH30658.12 SH30658.11 SH30662.14 SH30662.11 SH30657.14 SH30658.15 SH30658.16
82013-664PK 334.54 USD InStock
82013-664 82013-686 82013-674 82013-662 82013-684 82013-672 82013-660 82013-670 82013-680 82013-658 82013-668 82013-678 82013-688 82013-666 82013-654 82013-676
Labtainer™ BioProcess Containers, Thermo Scientific
Bags Media Bags
Small-volume, flexible BioProcess containers are ideal for process development applications.
  • Features two end dispensing lines that are capped or plugged
  • Bags arrive ready to use and have been sterilized via gamma irradiation
The standard 2-D hanging pillow containers are made available in medical-grade film. This animal-derived, component-free film is superior to others offered for biopharmaceutical and bioresearch applications due to outstanding tensile properties, low gas permeability, and low profile of leachable compounds.
These 2-D hanging pillow bags are completely disposable, which minimizes the risk of cross-contamination, eliminates cleaning, and reduces turnaround time in the lab. These containers are available in sizes ranging from 50 ml to 2 L and are suitable for packaging a variety of fluids including media, buffers, and process liquids.
Applications include QC sampling, product storage, and delivery and collection of cells. Small volume containers allow for complete fluid recovery and replace traditional rigid-walled plastic and glass containers, tubes, and flasks.
Ordering information: All container ports are equipped with tube extensions of varying length and LL (Luer-lock) or QC (quick connect) tubing connectors.
Packaging: Multipacks are individually poly-bagged, and poly-bagged per 10.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty