Would you like to log in again?
VWR® Floating Foam Tube Racks
Supplier: VWR
VWR®
HS2165AV HS2166V HS2165BV HS2165CV
82017-634CS 11414.4 USD InStock
82017-634 82017-632 82017-630 82017-628
VWR® Floating Foam Tube Racks
Racks Tube Racks
These high-density polyethylene foam racks are ideal for floating tubes in water baths, tubs, or beakers.
  • Resistant to water absorption
  • Color-coded for easy identification
  • Different shapes available
Closed cell foam is resistant to water absorption so the racks can be washed and reused. Foam handles make it easy to lift samples out of baths. Racks come in different colors and shapes for easy identification. Round rack 82017-634 is sized to fit a beaker and features unique x-shaped slits that accommodate a variety of tube sizes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty