Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
UVP MultiDoc-It™ Imagers, Analytik Jena
Supplier: Analytik Jena
MultiDoc-It™
97-0193-05 97-0202-02 97-0202-01 97-0197-05 97-0194-05 97-0195-04 97-0192-05 97-0192-04 97-0193-04 97-0195-05 97-0197-04
82020-696EA 8646.18 USD InStock
82020-696 82020-686 82020-700 82020-698 82020-688 82020-726 82020-692 82020-708 82020-728 82020-690 82020-706
UVP MultiDoc-It™ Imagers, Analytik Jena
Photodocumentation Systems
The UVP MultiDoc-It™ Imager is ideal for laboratories performing image acquisition of routine electrophoresis of DNA and protein gels.
  • System works with fluorescent and colorimetric gels, autoradiography film, and microtiter plates
The single intensity M-26 UVP Transilluminator operates on 115V, 8W, and offers a 302nm wavelength
It is supplied with a built-in 21x26cm (81/4x101/4"") ethidium bromide emission filter. The high resolution digital color camera has 12 megapixels, 640x480 to 4000x3000 resolution, 6x optical zoom, 4x digital zoom, and a USB connector. It fits securely into the topside chamber of the darkroom.
The compact metal darkroom enclosure has an overhead white LED light, and a wide access door with a UV-blocking viewing window; UV shuts off when door is open.
The image capture software controls camera functions including live image preview, focus and exposure settings, and image saturation warnings. It also features enhancement and annotation, detailed and configurable reporting, and history tracking.
Imaging of non-fluorescent gels requires UV to White converter plate 21474-892. UVP Visi-Blue™ Converter plate 15000-088 converts ultraviolet to 480nm blue light for use with SYBR® Green, SYPRO® Orange, and EGFP stains. Both plates measure 21x26cm (81/4x101/4"").
Certifications: CSA certified. CE compliant.
Ordering information: System includes transilluminator, filter, camera, darkroom, and VisionWorks Acquisition and Analysis Software. Additional UVP MultiDoc-It™ systems are available; contact your Avantor sales representative for more information.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty