Would you like to log in again?
Totally Tubular Tube Rack System, Neon Colors, USA Scientific
Supplier: USA Scientific
2344-1040 2344-1040
82020-732EA 247.66 USD InStock
82020-732 102952-538
Totally Tubular Tube Rack System, Neon Colors, USA Scientific
Racks Tube Racks
The Totally Tubular modular rack system includes one set of four tube blocks in mixed neon colors and one tray per pack.
  • Four removable tube blocks
  • Made of durable polypropylene
Modular rack includes four removable blocks that can be positioned to support four sizes of tubes. Durable polypropylene withstands freezing and autoclaving. Mixed neon colors include orange, green, pink, and yellow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty