Would you like to log in again?
VWR® Calibrated Electronic Thermometers with Stainless Steel Probe
Supplier: VWR
VWR®
60900-0411
82020-744EA 67.48 USD InStock
82020-744
VWR® Calibrated Electronic Thermometers with Stainless Steel Probe
Thermometers Digital Thermometers
Monitor the temperature of pharmaceuticals, vaccines, chemicals, blood and blood products, food products and more with these high accuracy digital thermometers with chemical resistant probes.
  • Features available include minimum/maximum temperature readings and timers
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
  • Individually serialized, CE marked
  • Aligned with GLOBE protocols
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty