Synonyms:
Dihydrogen oxide
SH30221.26
SH30221.26
SH30221.10
SH30221.25
SH30221.24
SH30221.28
SH30221.27
SH30221.24
SH30221.28
SH30221.27
SH30221.LS
SH30221.25
SH30221.17
16750-108PK
182.05
USD
InStock
16750-108
82013-254
CA82013-644
24125-501
CA82013-646
82007-324
CA16777-648
82020-796
CA82020-796
16777-648
82013-646
82013-644
CA82013-254
Water, HyClone™ HyPure for injection (WFI), sterile filtered
Water
Water for Injection (WFI) quality water supports cell culture growth and biomanufacturing processes.
- Meets stringent USP specifications
- Manufactured in ISO 9001 certified facility
- Lot-specific certificate of analysis (CoA) for each lot of water produced
- Intended for further manufacturing or research use. Not for diagnostic or therapeutic use
- 0.1 µm sterile filtered.
HyPure WFI Quality Water undergoes extensive purification to meet or exceed the stringent specifications of the United States Pharmacopeia (USP).
HyPure WFI Quality Water meets the following criteria: Must be produced from a validated water purification system that meets or exceeds current USP requirements for HyPure WFI Quality Water. Must have a quality assurance monitoring system that ensures continuous compliance and incorporates system test and action limits. Must meet all applicable chemical, purity, and other acceptance criteria as outlined in the current USP for both system and packaged water. Must include a lot-specific CoA for each lot of water produced, listing both acceptance criteria and actual test results. Actual test results on a CoA are not taken from system samples, but must be from actual packaged samples of the given lot.
Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Ordering information: All purified waters are supplied in PETE/PETG bottles, except for 82020-796, 82013-254, 16777-648, 82013-644, and 82013-646, each of which is supplied in a Thermo Scientific HyClone® BioProcess Container (BPC®) system. For alternate volumes and container options, contact your VWR sales representative.