Would you like to log in again?
Howard Leight Fusion™ Detectable Earplugs, Honeywell Safety
Supplier: Honeywell Safety
Howard Leight Fusion™
FDT-30-SM FDT-30
82020-866CS 2322.56 USD InStock
82020-866 82020-868
Howard Leight Fusion™ Detectable Earplugs, Honeywell Safety
Earplugs Reusable Earplugs
These multiple-use earplugs use advanced manufacturing technology to deliver a revolutionary combination of all-day comfort and easy handling.
  • FlexiFirm® core ensures easy insertion into the ear canal and a superior fit
  • SoftFlange™ design adjusts perfectly to the ear canal to deliver unmatched comfort and superior attentuation
  • Two sizes provide optimal fit for comfort and safety
  • Metalized cord and stem ring offer detection for process industries
  • Earplugs are stored in an included HearPack® plastic case
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty