Would you like to log in again?
VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Supplier: VWR
VWR® CryoPro®
PK-A3-64 04A3-49 PK-A3-81 PK-A1-VWR-03 04A2-VWR-03 04A3-25 04A1-VWR-02M 04A2-VWR-02D 04A1-04-GEN PK-A3-25 PK-A3-25M 04A1-02M-GEN 04A2-VWR-02 PK-A3-49 PK-A3-144 04A3-100 PK-A1-VWR-02 04A1-VWR0281 04A3-64 PK-A3-16 04A1-VWR-02 PK-A3-36 04A3-81 PK-A3-100 04A3-16
82007-146CS 500.65 USD InStock
82007-146 82007-144 82021-114 82021-116 82007-148 97014-322 82007-152 82007-150 89214-738 82021-122 82007-156 82021-124 89214-736 82007-154 82021-126 82007-158 82021-128 82021-120 82007-160 89214-740 82021-118 82007-142 89214-744 82007-162 89214-742
VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Boxes Cryoboxes
Boxes are constructed of durable fiberboard with a protective, moisture-repellant coating.
  • Available with or without slots or dividers
Liquid nitrogen (LN2) freezer boxes feature slots for safe LN2 drainage and reduced LN2 consumption. Mechanical freezer boxes do not feature slots and cannot be used with LN2. Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accomodate dividers (sold separately) for 16 to 100 cells. Fiberboard dividers can fit in any 13.2Wx13.2Lcm (51/5x51/5" ) LN2 cryogenic or mechanical freezer box.
Ordering information: Box dimensions include lid.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty