Would you like to log in again?
OrgoSol DetergentOUT™ Detergent Removal Systems, G-Biosciences
Supplier: G-Biosciences
OrgoSol DetergentOUT™
786-127 786-128 786-127 786-128
82021-286EA 285.82 USD InStock
82021-286 CA82021-286 82021-288 CA82021-288
OrgoSol DetergentOUT™ Detergent Removal Systems, G-Biosciences
Cleaning Agents Decontaminants
OrgoSol DetergentOUT™ is suitable for removal of detergents from protein solutions, including hydrophobic protein solutions
OrgoSol DetergentOUT™ first concentrates the protein solution through precipitation and then the detergent is extracted and removed with the supplied OrgoSol™ buffer. The proprietary precipitation agent ensures >99% protein recovery, however precipitation may result in some loss of a protein's biological activity.
Two sizes are offered: Micro Kit for processing up to a total of 10mL protein solution and Medi Kit for processing up to a total of 30mL protein solution, either in a single or multiple experiments.
OrgoSol DetergentOUT™ is compatible with all detergent types and its performance is not dependent on the concentration of detergents in the solution. OrgoSol DetergentOUT™ is highly flexible and can be used to process small and large sample volumes.
Ordering information: Accessories not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty