Would you like to log in again?
SDS Detection & Estimation Reagent Kit, G-Biosciences
Supplier: G-Biosciences
786-129
82021-290EA 166.47 USD InStock
82021-290 CA82021-290
SDS Detection & Estimation Reagent Kit, G-Biosciences
Protein Extraction Protein Extraction Kits
A reagent kit for detection and estimation of SDS in a sample
The working range of the assay is 0.02-0.2% SDS.
Mix the test sample in the extraction buffer reagents provided with the kit. If SDS is present in the sample, a blue color is extracted that can be quantitatively measured.
Ordering information: Accessories not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty