Would you like to log in again?
Protein dotMETRIC™ Assay, G-Biosciences
Supplier: G-Biosciences
786-21 786-20 786-20 786-21
82021-354EA 424.18 USD InStock
82021-354 CA82021-354 82021-360 CA82021-360
Protein dotMETRIC™ Assay, G-Biosciences
Assays Protein Assays
Rapidly mix 1µL protein sample with the supplied dilution buffer and apply 1µL of the solution to the test strip by point-of-contact capillary action
For increased reproducibility and test reliability, the dotMETRIC™ kits are offered with an optional Spot Application Device. The Spot Application Device allows application of samples using fixed volume (1µL) capillary tips and simplifies the task of applying the protein solution on the test strips by point-of-contact capillary action. The Spot Application Device simplifies the application of one or more samples and improves the reliability of results. The dotMETRIC™ assay is able to resist common laboratory agents such as Triton® X-100, Triton® X-114, Thesit®, Tween® 20, Nonidet® P-40, SDS, reducing agents such as ß-mercaptoethanol and DTT, sugars, cobalt, EDTA, Tris buffers, and so forth.
For rapid estimation of protein concentration in cellular fractions, tissue and cell lysates, and chromatography purification fractions.
Under the assay’s specific buffer conditions the protein enters into the matrix of the test strip, binds and saturates as protein then diffuses in a circular manner. Develop the test strip in approximately five minutes. A circular protein spot is produced. By measuring the diameter of the spots with the Protein dotMETRIC™ scale you can easily determine concentration of protein. No expensive spectrophotometers or cuvettes required.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty