Would you like to log in again?
LabSafe™ GEL Blue for Staining of Polyacrylamide Gels, G-Biosciences
Supplier: G-Biosciences
LabSafe™
786-35G 786-35 786-35 786-35G
82021-412EA 167.16 USD InStock
82021-412 82021-414 CA82021-412 CA82021-414
LabSafe™ GEL Blue for Staining of Polyacrylamide Gels, G-Biosciences
Stains Electrophoresis Stains
LabSafe GEL Blue™ is an enhanced protein stain based on Coomassie dye that offers unsurpassed sensitivity and rapid band visualisation.
  • Sensitive, 4-8ng BSA
  • Saves time, band develops within 3 minutes
  • Avoids the use of methanol, acetic acid, or other toxic agents
  • Works with native PAGE, SDS-PAGE, isoelectric focusing, or 2D gels
LabSafe GEL Blue™ is an ultra sensitive stain that is able to detect as little as 4ng protein. Protein bands are visible in as little as 3 minutes, with maximal staining achieved in an hour. The figure shows that LabSafe GEL Blue™ can detect protein levels as low as 4ng BSA. 60 to 1000ng BSA is detectable in 5 to 10 minutes, and the low levels of BSA (<8ng) become clearly visible when washed in water.
Works with native PAGE, SDS-PAGE, denaturing, isoelectric focusing, or 2D gel. Mass spectrometry compatible.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty