Would you like to log in again?
DNAOUT™ DNA Removal Solution, G-Biosciences
Supplier: G-Biosciences
786-74 786-75 786-75 786-74
82021-436EA 66.56 USD InStock
82021-436 CA82021-436 82021-438 CA82021-438
DNAOUT™ DNA Removal Solution, G-Biosciences
Cleaning Agents Decontaminants
DNAOUT™ is a DNA removal solution for the rapid decontamination of work areas from contaminating DNA. Simply spray the work area and wait 10-15 seconds and then rinse with Molecular Grade Water.
  • Ready-to-use with convenient spray bottle and refill bottles available
  • Remove contaminating DNA prior to sensitive work, including PCR
  • Removes amplicon, plasmid and genomic DNA
  • Fast acting (10-15 seconds)
  • Ideal for cleaning centrifuge tubes, reaction tubes, PCR machine surface, pipettes, lab benches, lab equipment and more
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty