Would you like to log in again?
Apoptotic DNA Ladder, G-Biosciences
Supplier: G-Biosciences
786-209
82021-578EA 410.65 USD InStock
82021-578 CA82021-578
Apoptotic DNA Ladder, G-Biosciences
Electrophoresis Reagents Molecular Weight Markers
G-Biosciences' Apoptotic DNA Ladder kit contains all of the reagents necessary for the preparation of cellular fractions for the production of apoptosis nucleosomal DNA ladders
The Apoptotic DNA Ladder kit protocol involves the following: cell lysis, removal of cellular debris, and precipitation of nucleosomal DNA. Nucleosomal DNA is detected after standard 1.8% agarose gel electrophoresis.
G-Biosciences' Apoptotic DNA Ladder protocol provides the choice of preparing nucleosomal DNA ladder either with or without genomic DNA. The kit is suitable for preparing up to 100 apoptotic ladders.
The kit does not require the use of toxic phenol
Ordering information: Accessories available
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty