Would you like to log in again?
CytoScan™ LDH Cytotoxicity Assay for Detection of Cell Toxicity, Death, Viability or Proliferation, G-Biosciences
Supplier: G-Biosciences
786-210
82021-582EA 616.97 USD InStock
CA82021-582 82021-582
CytoScan™ LDH Cytotoxicity Assay for Detection of Cell Toxicity, Death, Viability or Proliferation, G-Biosciences
Assays Cellular Assays
G-Biosciences' CytoScan™ LDH Cytotoxicity Assay is a colorimetric method of assaying cellular cytotoxicity and is an alternative to ⁵¹Cr release cytotoxicity assays
G-Biosciences' CytoScan™ LDH Cytotoxicity Assay Kit can be used with different cell types for assaying cell mediated cytotoxicity as well as cytotoxicity mediated by chemicals and other test compounds. The assay quantitatively measures the stable, cytosolic, lactate dehydrogenase (LDH) enzyme, which is released from damaged cells. The released LDH is measured with a coupled enzymatic reaction that results in the conversion of a tetrazolium salt (iodonitrotetrazolium (INT)) into a red color formazan by diaphorase.
The LDH activity is determined as NADH oxidation or INT reduction over a defined time period. The resulting formazan absorbs maximally at 492nm and can be measured quantitatively at 490nm using a micro-plate reader or spectrophotometer.
G-Biosciences' CytoScan™ LDH Cytotoxicity is supplied in a kit that contains components sufficient for performing up to 1000 assays.
The kit combines the advantages of reliability and simple evaluation, characteristic of radioisotope release assays, with the convenience of accuracy and avoidance of radioactivity.
Ordering information: Accessories available
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty