Would you like to log in again?
CytoScan™-fluoro Cytotoxicity Assay, G-Biosciences
Supplier: G-Biosciences
786-211 786-211
82021-584EA 607.11 USD InStock
82021-584 CA82021-584
CytoScan™-fluoro Cytotoxicity Assay, G-Biosciences
Assays Cellular Assays
G-Biosciences' CytoScan™-fluoro Cytotoxicity Assay, for the detection of cell toxicity, death, viability or proliferation, is ideal for high throughput screening.
G-Biosciences' CytoScan™-fluoro Cytotoxicity Assay is based on the quantification of cellular lactate dehydrogenase (LDH) released when cells are damaged or under stress. Lactate dehydrogenase released into the culture medium is measured with a diaphorase coupled enzymatic assay that results in the conversion of a non-fluorescent compound (resofurin) to a fluorescent compound (resorufin), which is measured using a fluorometer. The assay detects even low level damage to cell membrane not detected with other methods.
G-Biosciences' CytoScan™-fluoro Cytotoxicity Assay does not damage healthy cells and the assay can be performed directly in the cell culture wells containing a mixed population of viable and damaged cells. The kit is supplied with substrate mix, assay buffer, and stop solution.
G-Biosciences' CytoScan™-fluoro Cytotoxicity Assay is supplied in a kit that contains components sufficient for performing up to 500 assays.
Ordering information: Accessories available
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty