Would you like to log in again?
Arrest™ Extraction Buffer, A Chaotropic RNA Extraction Buffer, G-Biosciences
Supplier: G-Biosciences
Arrest™
786-133 786-133
82022-524EA 277.67 USD InStock
CA82022-524 82022-524
Arrest™ Extraction Buffer, A Chaotropic RNA Extraction Buffer, G-Biosciences
Nucleic Acid Reagents Transformation Reagents
G-Biosciences' Arrest™ Extraction Buffer is an optimized combination of various chaotropic agents and RNase inhibitors, which inhibits RNase in 5-10 minutes
G-Biosciences' Arrest™ Extraction Buffer has a quick, 10 minute, single step protocol that isolates high quality total RNA from most species and tissues. The protocol involves homogenization and lysis of samples in Arrest™ Extraction Buffer. After removing cellular debris, pure RNA is precipitated from the supernatant with ethanol.
G-Biosciences' Arrest™ Extraction Buffer is conveniently packaged as two 50 mL bottles; enough reagent for isolation of RNA from 10 grams of tissue or 200 preps of 50 mg tissue each.
Arrest™ Extraction Buffer may be used in conjunction with any RNA extraction method, including extractions based on phenol, chloroform and other organic solvents and detergents.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty