Would you like to log in again?
BLOK™ BLOTTO, 5% Modified Milk Protein Blocking Solution, G-Biosciences
Supplier: G-Biosciences
786-192
82022-630EA 206.11 USD InStock
CA82022-630 82022-630
BLOK™ BLOTTO, 5% Modified Milk Protein Blocking Solution, G-Biosciences
Buffers Electrophoresis Buffers Blocking Buffers
G-Biosciences' BLOK™ BLOTTO is a pre-formulated, convenient, ready-to-use blocking solution
G-Biosciences' BLOK™ BLOTTO contains 5% (w/v) nonfat powdered milk in 25mM Tris and 150mM NaCl, (pH 7.4), with 0.01% Antifoam A, supplied in ready-to-use form.
G-Biosciences' BLOK™ BLOTTO works in about 10 to 15 minutes, and is suitable for Western blots, dot blots, ELISA, immunohistochemistry and other immunochemical applications.
BLOK™ BLOTTO is a 5% novel modified nonfat milk protein solution.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty