Would you like to log in again?
FOCUS™ Signal Protein, G-Biosciences
Supplier: G-Biosciences
786-250
82022-678EA 358.9 USD InStock
CA82022-678 82022-678
FOCUS™ Signal Protein, G-Biosciences
Protein Extraction Protein Extraction Kits
Lipid rafts are membrane microdomains that are enriched in caveolin, cholesterol, glycolipids, sphingolipids and glycosyl-phosphatidylinositol
G-Biosciences' FOCUS™ Signal Proteins kit fractionates lipid raft localized proteins from other cellular proteins by employing non ionic detergents. Signal Protein Extraction Buffer is a proprietary formulation of non-ionic detergents designed to efficiently extract and remove soluble proteins, leaving lipid rafts containing signal proteins as a detergent insoluble fraction. The resulting rafts are then solubilized in FOCUS™ Protein Solubilization Buffer, a supplied 2D electrophoresis compatible buffer or a different buffer of choice.
G-Biosciences' FOCUS™ Signal Protein kit is designed for >50 preps, where a prep is one of the following: 100 mg mammalian tissue, 50μL wet animal cell pellet, 50μL wet yeast pellet, 50μL wet bacteria pellet, or 250 mg plant tissue.
Lipid rafts are also known as detergent-insoluble glycolipid-enriched complexes (GEMs) or DIGs. Many signaling proteins, including glycosylphosphatidylinositol (GPI)-anchored proteins, doubly-acylated tyrosine kinases of the Src family, and transmembrane proteins, are located in lipid rafts. Lipid raft localized proteins have been shown to be involved in intracellular membrane trafficking and signaling.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty