Would you like to log in again?
FOCUS™ Glycoprotein, G-Biosciences
Supplier: G-Biosciences
786-253
82022-684EA 527.71 USD InStock
CA82022-684 82022-684
FOCUS™ Glycoprotein, G-Biosciences
Protein Extraction Protein Extraction Kits
G-Biosciences' FOCUS™ Glycoprotein kit is based on lectin binding of specific glycoproteins with terminal α-D mannosyl and α-D glycosyl proteins.
G-Biosciences' FOCUS™ Glycoprotein kit isolates glycoproteins from complex biological solutions using spin columns that contain lectin (Concanavalin A) bound resin with the capacity to bind and immobilize ~5mg glycoproteins. Column bound glycoproteins are eluted with a set of three rapid elution buffers. The proprietary serial elution allows for faster elution compared to other glycoprotein purification systems.
The eluted proteins are suitable for 2D electrophoresis and isoelectric focusing.
FOCUS™ Glycoprotein is designed for 10 x 1.5 mg protein samples.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty