Would you like to log in again?
PhosphoQuant™, G-Biosciences
Supplier: G-Biosciences
PhosphoQuant™
786-256 786-256
82022-698EA 550.42 USD InStock
CA82022-698 82022-698
PhosphoQuant™, G-Biosciences
Protein Purification Protein Purification Kits
PhosphoQuant™ is specifically designed for quick and reliable determination of whether a purified protein is phosphorylated and the extent of phosphorylation
The assay is based on the alkaline hydrolysis of phosphates from seryl and threonyl residues in phosphoproteins and the subsequent quantification of the released phosphate with a Molybdate dye.
Ordering information: Accessories are available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty