Would you like to log in again?
Sulfo-EMCS (N-(ε-maleimidocaproyloxy) sulfosuccinimide ester)
Supplier: G-Biosciences
Warning
Synonyms: Sulfo EMCS
BC17 BC17
82022-802EA 395.81 USD InStock
CA82022-802 82022-802
Sulfo-EMCS (N-(ε-maleimidocaproyloxy) sulfosuccinimide ester)
Sulfo-EMCS (N-(ε-maleimidocaproyloxy) sulfosuccinimide ester)
N-(ε-Maleimidocaproyloxy) sulfo succinimide ester Heterobifunctional Cross Linker.
Order Now
Specification Test Results
Spacer Arm (Å) 9.4
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable NO
Water Soluble YES
Cleavable/ Reversible NO
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty