Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Photoreactive Cross Linkers, G-Biosciences
Supplier: G-Biosciences
BC35 BC39 BC33 BC28 BC30 BC29 BC28 BC38 BC34 BC32 BC38 BC29 BC35 BC32 BC30 BC39 BC33 BC34
82022-828EA 601.45 USD InStock
82022-828 CA82022-834 CA82022-824 82022-834 CA82022-832 CA82022-822 82022-824 CA82022-830 CA82022-842 82022-822 CA82022-820 82022-833 CA82022-828 82022-830 82022-820 CA82022-840 82022-842 82022-840
Photoreactive Cross Linkers, G-Biosciences
Chromatography Sorbents SPE Sorbents
G-Biosciences’ cross-linking agents contain at least two reactive groups that are reactive towards numerous groups, including sulfhydryls, amines and carbohydrates, and create chemical covalent bonds between two or more molecules. Functional groups that can be targeted with cross-linking agents are primary amines, carboxyls, sulfhydryls, carbohydrates and carboxylic acids. Protein molecules have many of these functional groups and therefore proteins and peptides can be readily conjugated using cross-linking agents.
G-Biosciences’ cross-linking agents are used for the following applications: protein structural and functional studies, cell membrane structural studies, protein & receptor interactions, cell surface studies, solid-phase immobilization, preparation of immunogens, immunotoxins and other conjugated protein reagents, protein to DNA/RNA cross-linking, reactive group transfer, protein-protein conjugation, and for immunological tools.
To select a cross-linking reagent several factors need to be considered (see selection guide). These factors include: Reactive Toward: determines the target residues to be cross-linked (select a reagent that does not interfere with protein function); Membrane Permeability: for cell surface labeling, select non-membrane permeable reagents; Cleavable: for easy release of cross-linked proteins from solid supports or for further downstream applications; Reversible: an alternative to cleavable reagents; Steric Hinderance: bulky groups around the binding site may require reagents with longer spacer arms.
G-Biosciences offers a variety of cross-linking agents, reaction specific buffers and accessories for performing and facilitating cross-linking applications. G-Biosciences’ cross-linking agents are divided into groups dependent on the number and similarity of the reactive groups. Photoreactive groups are used when no specific groups are available to react with as photoreactive groups react non-specifically upon exposure to UV light.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty