Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guanidinium hydrochloride

Supplier: G-Biosciences
Warning
Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideGuanadine hydrochloride

RC-057 BC88 BC88 BC85 RC-057 BC85
82022-848EA 268.17 USD InStock
CA71003-304 CA82022-848 82022-848 CA71003-410 71003-304 71003-410
Guanidinium hydrochloride
Guanidinium hydrochloride
A selection of strong chemicals to aid in the denaturation of proteins. Denaturants, or chaotropes, disrupt water interactions resulting in the solubilization of hydrophobic proteins and peptides. The chaotropes also act on all proteins to unfold them and alter their three-dimensional structure.

Available as a powder or 8M solution


Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty