Would you like to log in again?
Zwitterionic Detergent Solutions, G-Biosciences
Supplier: G-Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DG049 DG062 DG058 DG053 DG050 DG055 DG054 DG060 DG051 DG056 DG060 DG056 DG052 DG052 DG049 DG054 DG051 DG065 DG063 DG062 DG050 DG063 DG055 DG059 DG053 DG058 DG061 DG064 DG057 DG065 DG057 DG059 DG064
89168-106EA 70.85 USD InStock
89168-106 89168-108 82023-040 82023-024 CA82023-034 82023-026 82023-020 82023-064 82023-042 CA82023-030 95029-296 82023-022 95029-298 CA82023-032 CA95029-298 CA82023-026 82023-018 CA82023-028 CA89168-108 CA89168-106 82023-030 CA82023-022 CA82023-024 82023-016 CA82023-040 82023-032 CA95029-296 CA82023-042 CA82023-020 82023-034 82023-028 CA82023-016 CA82023-018
Zwitterionic Detergent Solutions, G-Biosciences
Cleaning Agents Detergents and Surface Cleansers
Zwitterionic detergents protect the native state of proteins without altering the native charge of the protein molecules
G-Biosciences Zwitterionic Detergent Solutions include: CHAPS, CHAPSO, Sulfobetaine 3-10 (SB 3-10), Sulfobetaine 3-12 (SB 3-12), Sulfobetaine 3-14 (SB 3-14), ASB-14, ASB-16, and ASB-C8Ø.
Zwitterionic detergents are used for isoelectric focusing and 2D electrophoresis. Synthetic zwitterionic detergents are known as sulfobetaines. Sulfobetaines retain their zwitterionic characteristics over a wide range of pH.
Ordering information: Accessories not included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty