Would you like to log in again?
Human Single Tissue Single Species Blot, Normal Tissue, G-Biosciences
Supplier: G-Biosciences
TB66 TB22 TB69 TB25 TB04 TB62 TB16 TB04 TB25 TB07 TB22 TB19 TB13 TB13 TB68 TB71 TB16 TB65 TB10 TB10 TB60 TB67 TB07 TB70 TB01 TB64 TB01 TB19 TB63 TB61
82023-160EA 456.79 USD InStock
82023-160 CA82023-154 CA82023-178 82023-148 95029-350 CA95029-360 CA82023-172 82023-142 CA95029-340 95029-352 CA82023-130 82023-166 CA95029-352 95029-344 CA95029-354 95029-346 CA95029-356 95029-348 CA82023-148 CA95029-358 CA95029-338 82023-172 CA82023-160 82023-178 CA82023-166 82023-136 95029-360 82023-130 95029-340 82023-154 CA95029-350 95029-342 CA82023-142 CA95029-342 95029-354 95029-356 CA95029-344 CA82023-136 95029-358 CA95029-346 CA95029-348 95029-338
Human Single Tissue Single Species Blot, Normal Tissue, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, human, single tissue blots from normal tissues.
  • Blots are ready to be blocked and probed with the antibodies of choice
  • Each blot contains two protein lanes, a protein marker lane and a lane with GenLysate™ sample
G-Biosciences premade western blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty