Would you like to log in again?
Rat Single Tissue Single Species Blot, G-Biosciences
Supplier: G-Biosciences
TB18 TB12 TB24 TB15 TB06 TB09 TB03 TB27 TB21
82023-170EA 456.79 USD InStock
82023-170 82023-182 CA82023-170 82023-140 CA82023-182 82023-134 CA82023-176 82023-146 82023-158 CA82023-134 CA82023-146 82023-152 82023-164 CA82023-140 82023-176 CA82023-152 CA82023-164 CA82023-158
Rat Single Tissue Single Species Blot, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, single tissue (normal) rat blots.
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty