Would you like to log in again?
Human Normal Cell Line Blots, G-Biosciences
Supplier: G-Biosciences
TB50 TB51 TB52 TB53
82023-234EA 1238.4 USD InStock
CA82023-236 CA82023-232 82023-234 CA82023-234 82023-236 82023-230 CA82023-230 82023-232
Human Normal Cell Line Blots, G-Biosciences
Blotting Kits
G-Biosciences offers ready-to-screen, normal tissue human cell line blots from epithelial, endothelial, skin, and muscle tissue.
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
Ordering information: Accessories available.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty