Would you like to log in again?
Protease Assay Kit, G-Biosciences
Supplier: G-Biosciences
786-028 786-028
82023-260EA 473.59 USD InStock
82023-260 CA82023-260
Protease Assay Kit, G-Biosciences
Assays Protein Assays
The Protease Assay Kit is designed for the quantitative determination of proteases present in protein sample, using a dye-labeled protein substrate
Chemically stabilized Trypsin (MSG-Trypsin™) is supplied with the kit as a general protease standard; however, other specific protease standards can also be used. MSG-Trypsin™ is an ultra-pure trypsin from bovine pancreas, modified by methylation followed by TPCK treatment and is extremely resistant to autolysis. The kit components are sufficient for 50 assays in a microtiter plate format or 0.5ml assay tubes.
Accessories: Resorufin-Casein Protease Substrate (#786-321): A colorimetric substrate that when treated with proteases releases resorufin that has an absorbance of 570nm. Supplied lyophilized. FITC-Casein Protease Substrate (#786-322): A fluorescent (fluorescein isothiocyanate (FITC)) substrate that when treated with proteases releases FITC that has an excitation at 485nm and emission at 535nm. Supplied lyophilized.
The proteases present in the sample of interest will digest the protein substrate and release dye labeled peptides. The absorbance of the dye-labeled peptide is measured at 570nm for determination of protease activity.
Ordering information: Accessories not included.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty