Would you like to log in again?
Biotin Conjugation Estimation, G-Biosciences
Supplier: G-Biosciences
BKC-01
82023-262EA 297.88 USD InStock
82023-262 CA82023-262
Biotin Conjugation Estimation, G-Biosciences
Protein Linking and Labeling Systems
G-Biosciences' HOOK™ BiotinQuant is designed for the estimation of biotin conjugation.
G-Biosciences' HOOK™ BiotinQuant™ uses HABA [4’-hydroxyazobenzene-2-carboxylic acid] dye to rapidly estimate the mole-to-mole ratio of biotin to antibody or protein. The method of biotin incorporation estimation is based on the binding of avidin with HABA dye, which produces a color that can be read at 500 nm. The HABA-avidin complex can be displaced with free biotin or biotin conjugated with other molecules (proteins). Measuring the change in optical density of HABA-avidin complex with biotinylated proteins allows for accurate estimation of the molar ration of biotin conjugated to the protein/ antibody.
G-Biosciences' HOOK™ BiotinQuant™ is supplied with premixed, OneQuant™ HABA/Avidin aliquots to eliminate difficulties in weighing small amounts of reagents and ensuring reproducible estimations.
Ordering information: Accessories included. Additional supplies available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty