Would you like to log in again?
Glycoprotein Staining Kit, G-Biosciences
Supplier: G-Biosciences
786-254 786-254
82023-286EA 439.98 USD InStock
CA82023-286 82023-286
Glycoprotein Staining Kit, G-Biosciences
Stains Electrophoresis Stains
G-Biosciences offers the Glycoprotein Staining kit for the highly sensitive detection of glycoproteins following gel electrophoresis or protein transfer to nitrocellulose membranes
G-Biosciences' Glycoprotein Staining kit uses an enhanced Periodic Acid-Schiff (PAS) method for detection of glycoprotein sugars. The supplied oxidizing agent first oxidizes the cis-diol sugar groups to aldehydes. The aldehyde groups react with the sensitive Glyco-Stain Solution forming Schiff bonds and producing strong magenta color bands.
G-Biosciences' Glycoprotein Staining kit is supplied with an enhanced Coomassie stain, called RAPIDstain™, that enhances the glycoprotein staining. RAPIDstain™ can also be used after glycoprotein staining to detect non-glycosylated proteins. The kit is sufficient for 10 mini gels (8 x 8 cm) or 20 nitrocellulose membrane (8 x 8 cm).
The Glycoprotein Staining kit is highly convenient as all the key reagents required for staining are supplied and a unique positive & negative control is included. In addition, the kit allows for the detection of glycosylated and non-glycosylated proteins on a single gel or membrane.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty