Would you like to log in again?
SmartWall Plus™ Storage System, Metro™
Supplier: Metro International
SmartWall Plus™ SmartWall®
1230CSNC 1230CSNC
82023-314EA 106.67 USD InStock
82023-314 CA82023-314
SmartWall Plus™ Storage System, Metro™
Furniture Shelves
Designed to optimize wall storage through a variety of shelf, accessory grid, and cabinet combinations organized onto easy-to-install wall tracks.
  • Corrosion-proof Type 304 stainless steel components
  • Cantilever shelves attach directly to posts without shelf supports
  • Taupe post assemblies including steel brackets are coated in corrosion-resistant taupe epoxy
Corrosion-resistant taupe epoxy components address corrosion and durability issues and in a multitude of applications. Wall tracks are available in 16 gauge Type 304 stainless steel and corrosion-resistant taupe epoxy finishes. Holes on wall tracks are spaced 20.3cm (8") apart; first and last holes are positioned 10.2cm (4") from track ends. Weight capacities per unit depend on the method of mounting and wall structure. Joining braces must be used to connect adjacent tracks. Post assemblies must be secured to wall surfaces through the bottom bracket. Smartwall Plus™ is only to be used with shelves that are 45.7cm (18") deep or less. Brackets for chrome, Metroseal 3, and stainless steel post assemblies are Type 304 stainless steel.
Ordering information: Wall tracks are supplied with two aluminum joining braces/stops, two self-tapping stainless steel screws for joining braces, and two plastic end caps. Post assemblies include top bracket, intermediate bracket, bottom bracket, standard Super Erecta® post, and stainless steel bolt for bottom bracket. For additional SmartWall Plus™ storage and accessory items, see 83009-610 series. For easy-order starter kits, see 82023-520 series. Super Erecta® shelves require shelf supports, two single supports per shelf. Double shelf supports are intended for a run of multiple shelves. Super Erecta® shelves are available in additional finishes (see 19766-070, 19766-072, and 83009-332 series). MetroMax™ corner adapters are required to allow MetroMax™ shelves to be used with standard shelf supports.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty