Would you like to log in again?
HD qwikTRAK™ High-Density Heavy-Duty Storage System, Metro™
Supplier: Metro International
qwikTRAK™
BTS7NA BTES BTS6NA BTEK3 BTS6NA BTS14NA BTAK3 BTS20NA BTMC BTAS BTS17NA BTMK3 BTS18NA BTS14NA BTMS BTS12NA BTS15NA BTAC BTS20NA BTAS BTS11NA BTS17NA BTMS BTS9NA BTS10NA BTEC BTS18NA BTS9NA BTS19NA BTS19NA BTS11NA BTS16NA BTEK3 BTS8NA BTAC BTS13NA BTS21NA BTS16NA BTS8NA BTEC BTS7NA BTAK3 BTMK3 BTS10NA BTS13NA BTS15NA BTES BTMC BTS12NA BTS21NA
82023-380EA 754.88 USD InStock
82023-380 CA82023-390 82023-382 CA82023-396 82023-420 82023-388 CA82023-398 CA82023-392 82023-384 CA82023-394 82023-386 82023-418 82023-414 82023-416 82023-390 82023-392 82023-398 82023-394 82023-396 CA82023-374 82023-400 CA82023-376 CA82023-410 CA82023-372 CA82023-404 CA82023-406 CA82023-400 CA82023-402 CA82023-408 CA82023-380 82023-376 82023-410 CA82023-386 82023-378 CA82023-420 82023-412 CA82023-388 82023-372 CA82023-382 82023-374 CA82023-384 82023-406 CA82023-416 82023-408 CA82023-418 CA82023-378 82023-402 CA82023-412 82023-404 CA82023-414
HD qwikTRAK™ High-Density Heavy-Duty Storage System, Metro™
Furniture Shelves
Designed to maximize storage capacity while increasing accessibility of shelved items.
  • Can increase storage capacity of a given area by 30 to 40%
  • System is composed of stationary units, mobile units, and a guiding track
Stationary end unit kits include the hardware necessary for connecting end units to the track and anchoring the track to floor surfaces. One kit is required for the entire system. Stationary intermediate unit kits also include the appropriate hardware for connecting ends to the track. One kit is required for the entire system. Mobile unit kits include casters, donut bumpers, and the necessary hardware. All end, intermediate, and mobile unit kits include 188 cm (74") high posts, and can hold up to 907 kg (2000 lbs.). Track sets offer a variety of sections of track for assembling a track run ranging from 1.8 to 6.1 m (6 to 21'). Track and joining channel are made of aluminum extrusion 6063 with 6.4 mm (¹/₄") stainless steel tubing. Joining kit is stainless steel. Track sets are supplied with 8.3 cm (3¹/₄") acetyl wheel casters.
Stationary units anchor the system and track into position. Mobile units are positioned between stationary units and permit a single aisle to be opened between any two units as needed, providing easy access to all stored products. The guiding track is assembled and positioned on the floor to provide a smooth and level surface for the mobile units.
Ordering information: HD qwikTRAK™ units are designed for use with Super Erecta® shelving and accessories. Shelves must be purchased separately and must be 45.7 cm (18") or wider. Shelves are available in chrome, stainless steel, or Metroseal 3 with Microban® built-in product protection (see 19766-070, 19766-072, and 83009-332 series).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty