Designed to maximize storage capacity while increasing accessibility of shelved items.
- Can increase storage capacity of a given area by 30 to 40%
- System is composed of stationary units, mobile units, and a guiding track
Stationary end unit kits include the hardware necessary for connecting end units to the track and anchoring the track to floor surfaces. One kit is required for the entire system. Stationary intermediate unit kits also include the appropriate hardware for connecting ends to the track. One kit is required for the entire system. Mobile unit kits include casters, donut bumpers, and the necessary hardware. All end, intermediate, and mobile unit kits include 188 cm (74") high posts, and can hold up to 907 kg (2000 lbs.). Track sets offer a variety of sections of track for assembling a track run ranging from 1.8 to 6.1 m (6 to 21'). Track and joining channel are made of aluminum extrusion 6063 with 6.4 mm (¹/₄") stainless steel tubing. Joining kit is stainless steel. Track sets are supplied with 8.3 cm (3¹/₄") acetyl wheel casters.
Stationary units anchor the system and track into position. Mobile units are positioned between stationary units and permit a single aisle to be opened between any two units as needed, providing easy access to all stored products. The guiding track is assembled and positioned on the floor to provide a smooth and level surface for the mobile units.
Ordering information: HD qwikTRAK™ units are designed for use with Super Erecta® shelving and accessories. Shelves must be purchased separately and must be 45.7 cm (18") or wider. Shelves are available in chrome, stainless steel, or Metroseal 3 with Microban® built-in product protection (see 19766-070, 19766-072, and 83009-332 series).